missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-ribose, 99%

CAS: 533-67-5 | C5H10O4 | 134.13 g/mol

$444.22 - $444.22

Chemical Identifiers

CAS 533-67-5
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
MDL Number MFCD00135904
InChI Key PDWIQYODPROSQH-GLULIZFNNA-N
Synonym 2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
PubChem CID 5460005
ChEBI CHEBI:28816
IUPAC Name (3S,4R)-3,4,5-trihydroxypentanal
SMILES OC[C@H]1OC(O)C[C@@H]1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC153280250
SDS View Documents
Thermo Scientific Chemicals
153280250
25 g Glass bottle
Each for $444.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

533-67-5
134.13
PDWIQYODPROSQH-GLULIZFNNA-N
5460005
(3S,4R)-3,4,5-trihydroxypentanal
C5H10O4
MFCD00135904
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
CHEBI:28816
OC[C@H]1OC(O)C[C@@H]1O

Specifications

533-67-5
100.0
White to Yellow
1.0% max. (40°C, 2 hrs) (vacuum)
98.5% min
C5H10O4
25 g
-57
Solubility in water: soluble. Other solubilities: soluble in pyridine,slightly soluble in alcohol
OC[C@H]1OC(O)C[C@@H]1O
134.13
CHEBI:28816
99%
2-Deoxy-D-ribose, 99%
98.5
91°C
0.5% max.
Authentic
Glass bottle
MFCD00135904
15, 2909
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose
PDWIQYODPROSQH-GLULIZFNNA-N
(3S,4R)-3,4,5-trihydroxypentanal
5460005
134.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-573-0

RTECSNumber : SB7230000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only