missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-glucose, 98+%, Thermo Scientific Chemicals

CAS: 154-17-6 | C6H12O5 | 164.16 g/mol

Supplier:  Thermo Scientific Chemicals 111980050

Quantity 5 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC111980050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

154-17-6
164.16
PMMURAAUARKVCB-PHUJZJCSNA-N
17751002
OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O
C6H12O5
MFCD00151328
deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
(3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal

Specifications

2-Deoxy-D-glucose
98.5
146.0°C to 147.0°C
1% max.
Authentic
Glass bottle
MFCD00151328
15,2905
deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
PMMURAAUARKVCB-PHUJZJCSNA-N
(3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal
17751002
99%
154-17-6
100.0
White to Cream
1% max.
99%
C6H12O5
5 g
+ 45.50
(2% in water) Clear colorless to light yellow
OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O
164.16
164.16
Crystalline Powder or Crystals
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-823-

SDS
Documents

Documents

Promotions

Promotions