missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-glucose, 98+%, Thermo Scientific Chemicals

Catalog No. p-4258923
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111980010 1 g Glass bottle
AC111980050 5 g Glass bottle
AC111980250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111980010 Supplier Thermo Scientific Chemicals Supplier No. 111980010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 154-17-6 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154-17-6
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00151328
InChI Key PMMURAAUARKVCB-PHUJZJCSNA-N
Synonym deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
PubChem CID 17751002
IUPAC Name (3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 146.0°C to 147.0°C
Color White to Cream
Ignition Residue 1% max.
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,2905
Specific Rotation + 45.50
Solubility Information (2% in water) Clear colorless to light yellow
Formula Weight 164.16
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Deoxy-D-glucose
Show More Show Less
EINECSNumber 205-823-

RUO – Research Use Only