missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-glucose, 98%

CAS: 154-17-6 | C6H12O5 | 164.16 g/mol

Supplier:  Thermo Scientific Chemicals L0733806

 View more versions of this product

Catalog No. AAL0733806


Only null left

Description

Description

2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
Specifications

Specifications

2-Deoxy-D-glucose
147°C to 152°C
MFCD00151328
1723331
deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
PMMURAAUARKVCB-PHUJZJCSNA-N
OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O
164.16
164.16
154-17-6
C6H12O5
5 g
14,2904
Soluble in water.
+47° (c=0.5 in Water, 45h)
(3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal
17751002
98%

Chemical Identifiers

154-17-6
164.16
PMMURAAUARKVCB-PHUJZJCSNA-N
17751002
OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O
C6H12O5
MFCD00151328
deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
(3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-823-0

RTECSNumber : MQ3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only