Learn More
Thermo Scientific Chemicals 2-Deoxy-D-glucose, 98%
CAS: 154-17-6 | C6H12O5 | 164.16 g/mol
$125.00 - $1,577.30
Chemical Identifiers
| CAS | 154-17-6 |
|---|---|
| Molecular Formula | C6H12O5 |
| Molecular Weight (g/mol) | 164.16 |
| MDL Number | MFCD00151328 |
| InChI Key | PMMURAAUARKVCB-PHUJZJCSNA-N |
| Synonym | deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose |
| PubChem CID | 17751002 |
| IUPAC Name | (3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal |
| SMILES | OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL0733803
|
Thermo Scientific Chemicals
L0733803 |
1 g |
Each for $125.00
|
|
|||||
|
AAL0733806
|
Thermo Scientific Chemicals
L0733806 |
5 g |
Each for $456.14
|
|
|||||
|
AAL0733814
|
Thermo Scientific Chemicals
L0733814 |
25 g |
Each for $1,577.30
|
|
|||||
Description
2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Deoxy-D-glucose is act as a culture media part in molecular genetics and as a targeted optical imaging agent for fluorescent imaging. It finds an application in glucoprivic feeding research to invoke and study the processes of counter-regulatory response (CRR). It is utilized in the development of anti-cancer approaches like oxidative stress, radio and chemosensitization.
Solubility
Soluble in water.
Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
Chemical Identifiers
| C6H12O5 | |
| MFCD00151328 | |
| deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose | |
| (3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal |
Specifications
| 154-17-6 | |
| C6H12O5 | |
| 1723331 | |
| deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose | |
| PMMURAAUARKVCB-PHUJZJCSNA-N | |
| +47° (c=0.5 in Water, 45h) | |
| 164.16 | |
| 164.16 | |
| 2-Deoxy-D-glucose |
| 147°C to 152°C | |
| MFCD00151328 | |
| 14,2904 | |
| Soluble in water. | |
| OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O | |
| (3S,4R,5S)-3,4,5,6-tetrahydroxyhexanal | |
| 17751002 | |
| 98% |
Safety and Handling
EINECSNumber : 205-823-0
RTECSNumber : MQ3325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only