missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-galactose, 99%

For studying galactose uptake into Escherichia coli | CAS: 1949-89-9 | C6H12O5 | 164.16 g/mol

Supplier:  Thermo Scientific Chemicals J6548209

 View more versions of this product

Catalog No. AAJ6548209


Only null left

Description

Description

2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.
Specifications

Specifications

2-Deoxy-D-galactose
107°C to 110°C
C6H12O5
10 g
2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PMMURAAUARKVCB-ONLQHTGVNA-N
(3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal
102191
164.16
Powder
1949-89-9
White
MFCD00014649
1723333
Soluble in water.
OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
164.16
CHEBI:27411
99%

Chemical Identifiers

1949-89-9
164.16
PMMURAAUARKVCB-ONLQHTGVNA-N
102191
(3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal
C6H12O5
MFCD00014649
2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
CHEBI:27411
OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-765-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only