missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-galactose, 99%

Catalog No. p-7043089
Change view
Click to view available options
Quantity:
2 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6548204 2 g
AAJ6548206 5 g
AAJ6548209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6548204 Supplier Thermo Scientific Chemicals Supplier No. J6548204
Only null left

For studying galactose uptake into Escherichia coli | CAS: 1949-89-9 | C6H12O5 | 164.16 g/mol

2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Deoxy-D-galactose is used as an inhibitor of fucosylation, which is a process of adding hexose deoxy sugar units to a molecule. It is used for studying galactose uptake into Escherichia coli and also for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Provide appropriate exhaust ventilation at places where dust is formed.

Chemical Identifiers

CAS 1949-89-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00014649
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PubChem CID 102191
ChEBI CHEBI:27411
IUPAC Name (3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O

Specifications

Melting Point 107°C to 110°C
Color White
Quantity 2 g
Beilstein 1723333
Solubility Information Soluble in water.
Formula Weight 164.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Deoxy-D-galactose
EINECSNumber 217-765-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only