missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Deoxy-5-methylcytidine 98.0+%, TCI America™
$119.61 - $2358.08
Chemical Identifiers
| CAS | 838-07-3 |
|---|---|
| Molecular Formula | C10H15N3O4 |
| Molecular Weight (g/mol) | 241.247 |
| MDL Number | MFCD00006549 |
| InChI Key | LUCHPKXVUGJYGU-XLPZGREQSA-N |
| Synonym | 5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference |
| PubChem CID | 440055 |
| ChEBI | CHEBI:47876 |
| IUPAC Name | 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one |
| SMILES | CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D3610100MG
|
TCI America
D3610100MG |
100 mg |
Each for $119.61
|
|
|||||
|
D3610500MG
|
TCI America
D3610500MG |
500 mg |
Each for $343.66
|
|
|||||
|
D36105G
|
TCI America
D36105G |
5 g |
Each for $2,358.08
|
|
|||||
Chemical Identifiers
| 838-07-3 | |
| 241.247 | |
| LUCHPKXVUGJYGU-XLPZGREQSA-N | |
| 440055 | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one |
| C10H15N3O4 | |
| MFCD00006549 | |
| 5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference | |
| CHEBI:47876 | |
| CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O |
Specifications
| 838-07-3 | |
| White | |
| MFCD00006549 | |
| 5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference | |
| CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O | |
| 241.247 | |
| CHEBI:47876 | |
| ≥98.0% (HPLC,T) | |
| 2′-Deoxy-5-methylcytidine |
| 193°C | |
| C10H15N3O4 | |
| 100 mg | |
| LUCHPKXVUGJYGU-XLPZGREQSA-N | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one | |
| 440055 | |
| 241.25 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : HA3860000
TSCA : No