missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxy-5-methylcytidine 98.0+%, TCI America™

Supplier:  TCI America D3610500MG

 View more versions of this product

Catalog No. D3610500MG


Only null left

Description

Description

Specifications

Specifications

2′-Deoxy-5-methylcytidine
193°C
C10H15N3O4
500 mg
LUCHPKXVUGJYGU-XLPZGREQSA-N
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one
440055
241.25
Crystalline Powder
838-07-3
White
MFCD00006549
5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference
CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O
241.247
CHEBI:47876
≥98.0% (HPLC,T)

Chemical Identifiers

838-07-3
241.247
LUCHPKXVUGJYGU-XLPZGREQSA-N
440055
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one
C10H15N3O4
MFCD00006549
5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference
CHEBI:47876
CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : HA3860000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions