missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxy-5-fluorouridine 98.0+%, TCI America™

Supplier:  TCI America D22351G

Quantity 1 g

 View more versions of this product

Catalog No. D22351G


Only null left

Description

Description

Specifications

Chemical Identifiers

50-91-9
246.19
ODKNJVUHOIMIIZ-RRKCRQDMSA-N
5790
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
C9H11FN2O5
MFCD00006530
floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
CHEBI:60761
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

2′-Deoxy-5-fluorouridine
150°C
C9H11FN2O5
MFCD00006530
floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O
246.19
CHEBI:60761
≥98.0% (HPLC,T)
50-91-9
White
1 g
2811
ODKNJVUHOIMIIZ-RRKCRQDMSA-N
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
5790
246.19
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YU7525000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions