missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Désoxy-5-fluorouridine 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 150°C |
| Color | Blanc |
| Quantity | 100 mg |
| UN Number | 2811 |
| Formula Weight | 246.19 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 2′-Deoxy-5-fluorouridine |
Sécurité et manipulation
| RTECSNumber | YU7525000 |
| TSCA | No |