missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxy-5-fluorocytidine 98.0+%, TCI America™

Catalog No. D36425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36425G 5 g
D36421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36425G Supplier TCI America Supplier No. D36425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10356-76-0
Molecular Formula C9H12FN3O4
Molecular Weight (g/mol) 245.21
MDL Number MFCD00077348
InChI Key IDYKCXHJJGMAEV-UHFFFAOYNA-N
PubChem CID 515328
IUPAC Name 4-amino-5-fluoro-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1F)C1CC(O)C(CO)O1

Specifications

Melting Point 196°C
Color White
Quantity 5 g
Formula Weight 245.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxy-5-fluorocytidine
RTECSNumber HA3850000
TSCA No