accessibility menu, dialog, popup

UserName

CAS RN 10356-76-0

IUPAC Name: 4-amino-5-fluoro-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: 5-fluoro-2'-deoxycytidine
Molecular Weight (g/mol): 245.21
Molecular Formula: C9H12FN3O4
InChi Key: IDYKCXHJJGMAEV-UHFFFAOYNA-N
SMILES: NC1=NC(=O)N(C=C1F)C1CC(O)C(CO)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results