missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxy-2'-fluorocytidine Hydrate 98.0+%, TCI America™

Catalog No. D36141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D36141G Supplier TCI America Supplier No. D36141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10212-20-1
Molecular Formula C9H12FN3O4
Molecular Weight (g/mol) 245.21
MDL Number MFCD00057445
InChI Key NVZFZMCNALTPBY-XVFCMESISA-N
Synonym 2'-deoxy-2'-fluorocytidine, 2'-fluoro-2'-deoxycytidine, 4-amino-1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl pyrimidin-2-one, cytidine, 2'-deoxy-2'-fluoro, unii-lcy080jpy9, 2-deoxy-2-fluorocytidine, lcy080jpy9, 2'-fc, 2'-fluoro-d-cytidine
PubChem CID 101507
IUPAC Name 4-amino-1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)F

Specifications

Melting Point 167°C
Color White
Quantity 1 g
Formula Weight 245.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxy-2′-fluorocytidine Hydrate