accessibility menu, dialog, popup

UserName

NH2NONOOHHOF

CAS RN 10212-20-1

IUPAC Name: 4-amino-1-[3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: 4-amino-1-[3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Molecular Weight (g/mol): 245.21
Molecular Formula: C9H12FN3O4
InChi Key: NVZFZMCNALTPBY-UHFFFAOYNA-N
SMILES: NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1F
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)