missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Deoxy-2,2-difluoro-D-erythro-pentonic Acid gamma-Lactone 3,5-Dibenzoate 98.0+%, TCI America™
$29.83 - $78.29
Chemical Identifiers
| CAS | 122111-01-7 |
|---|---|
| Molecular Formula | C19H14F2O6 |
| Molecular Weight (g/mol) | 376.312 |
| MDL Number | MFCD08458308 |
| InChI Key | SHHNEUNVMZNOID-HUUCEWRRSA-N |
| PubChem CID | 9821015 |
| IUPAC Name | [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate |
| SMILES | C1=CC=C(C=C1)C(=O)OCC2C(C(C(=O)O2)(F)F)OC(=O)C3=CC=CC=C3 |
Chemical Identifiers
| 122111-01-7 | |
| 376.312 | |
| SHHNEUNVMZNOID-HUUCEWRRSA-N | |
| [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate |
| C19H14F2O6 | |
| MFCD08458308 | |
| 9821015 | |
| C1=CC=C(C=C1)C(=O)OCC2C(C(C(=O)O2)(F)F)OC(=O)C3=CC=CC=C3 |
Specifications
| 122111-01-7 | |
| White | |
| MFCD08458308 | |
| SHHNEUNVMZNOID-HUUCEWRRSA-N | |
| [(2R,3R)-3-benzoyloxy-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate | |
| 9821015 | |
| ≥98.0% (HPLC) | |
| 2-Deoxy-2,2-difluoro-D-erythro-pentonic Acid gamma-Lactone 3,5-Dibenzoate |
| 123°C | |
| C19H14F2O6 | |
| 1 g | |
| C1=CC=C(C=C1)C(=O)OCC2C(C(C(=O)O2)(F)F)OC(=O)C3=CC=CC=C3 | |
| 376.312 | |
| 376.31 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer