missing translation for 'onlineSavingsMsg'
Learn More

2-Decanone, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019497
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019497 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019497 Supplier Merck Emd Millipore Supplier No. 682285ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Decanone can be obtained via liquid-phase Wacker oxidation of 1-decene in the presence of an efficient catalyst like Pd60 (Pd- palladium) nanocluster immobilized on titanium oxide support and cuprous chloride/H2O.

Specifications

CAS 693-54-9
Vapor Density >1 (vs. air)
Density 0.825 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3(CH2)7 COCH3
MDL Number MFCD00009571
Melting Point 3.5°C
Boiling Point 211°C
Synonym Methyl octyl ketone
Quantity 5 mL
Formula Weight 156.27
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less