missing translation for 'onlineSavingsMsg'
Learn More

2-Decanone, 97%

Catalog No. AAA1414506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1414506 5 g
AAA1414514 25 g
AAA1414522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1414506 Supplier Thermo Scientific Chemicals Supplier No. A1414506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 693-54-9 | C10H20O | 156.269 g/mol

2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 693-54-9
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00009571
InChI Key ZAJNGDIORYACQU-UHFFFAOYSA-N
Synonym 2-decanone, methyl octyl ketone, methyl n-octyl ketone, octyl methyl ketone, unii-gx543olt0r, n-c8h17coch3, gx543olt0r, chembl47127
PubChem CID 12741
ChEBI CHEBI:77929
IUPAC Name decan-2-one
SMILES CCCCCCCCC(=O)C

Specifications

Melting Point 3°C
Density 0.824
Boiling Point 210°C to 212°C
Flash Point 86°C (187°F)
Refractive Index 1.425
Quantity 5 g
Beilstein 1747463
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 156.27
Percent Purity 97%
Chemical Name or Material 2-Decanone
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 211-752-6
RTECSNumber HE0725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only