missing translation for 'onlineSavingsMsg'
Learn More

Cytiva 2-D Quant Kit

Catalog No. 45001732
Change view
Click to view available options
Quantity:
500 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001732 500 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45001732 Supplier Cytiva Supplier No. 80648356
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Precipitant, 250mL; Co-precipitant, 250mL; Copper solution, 50mL; Color reagent A, 2 x 250mL; Color reagent B, 5mL; BSA standard (2mg/mL), 5mL

Use for accurate determination of protein concentration in samples prepared for electrophoresis techniques. Cytiva Life Sciences™ 2-D Quant Kit precipitates proteins while leaving interfering substances behind.

Designed for the accurate determination of protein concentration in samples prepared for electrophoresis techniques. Works by quantitatively precipitating proteins while leaving interfering substances behind. The assay is based on the specific binding of copper ions to protein.

  • Works in the presence of 2% SDS, 1% DTT, 8M urea, 2M thiourea, 4% CHAPS, 2% Pharmalyte™, and 2% IPG Buffer
  • Quantitatively precipitates proteins while leaving interfering substances behind
  • Linear response in the range of 0 to 50μg protein, with sample volumes of 1 to 50μL
  • Assay time: one hour

2-D electrophoresis, SDS-PAGE, IEF

Specifications

For Use With (Application) 2D Electrophoresis, SDS-PAGE, IEF
Includes 250mL Precipitant, 250mL Co Precipitant, 50mL Copper Solution, 2 x 250mL Color reagent A, 5mL Color reagent B, 5mL BSA standard (2mg/mL)
Sample Volume 1 to 50μL
Content And Storage 4° to 8°C
Product Type 2D Quant kit
Quantity 500 Assays