missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopropylmethoxy-3-iodopyridine, 97%

Catalog No. p-7033074
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5008403 1 g
AAH5008406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5008403 Supplier Thermo Scientific Chemicals Supplier No. H5008403
Only null left

CAS: 766557-62-4 | C9H10INO | 275.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766557-62-4
Molecular Formula C9H10INO
Molecular Weight (g/mol) 275.09
MDL Number MFCD06659010
InChI Key ILSBVXOYWKUWLJ-UHFFFAOYSA-N
Synonym 2-cyclopropylmethoxy-3-iodo-pyridine, 2-cyclopropylmethoxy-3-iodopyridine, 2-cyclopropyl-methoxy-3-iodopyridine, pubchem16562, acmc-20anx8, pyridine,2-cyclopropylmethoxy-3-iodo, 2-cyclopropylmethoxy-3-iodanyl-pyridine, pyridine, 2-cyclopropylmethoxy-3-iodo
PubChem CID 17750186
IUPAC Name 2-(cyclopropylmethoxy)-3-iodopyridine
SMILES IC1=CC=CN=C1OCC1CC1

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 275.09
Percent Purity 97%
Chemical Name or Material 2-Cyclopropylmethoxy-3-iodopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only