missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopropyl-L-glycine, 97%

Catalog No. p-7056684
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH65755MD 250 mg
AAH6575503 1 g
AAH6575506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH65755MD Supplier Thermo Scientific Chemicals Supplier No. H65755MD
Only null left

CAS: 49606-99-7 | C5H9NO2 | 115.13 g/mol

2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopropyl-L-glycine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Store at 4°C. Incompatible with heat and oxidizing agents. Protect from heat.

Chemical Identifiers

CAS 49606-99-7
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD06659116
InChI Key BUSBCPMSNBMUMT-BYPYZUCNSA-N
Synonym l-cyclopropylglycine, s-2-amino-2-cyclopropylacetic acid, h-cyclopropyl-gly-oh, 2-cyclopropyl-l-glycine, l-cyclopropyl glycine, 2s-2-amino-2-cyclopropylacetic acid, s-amino-cyclopropyl-acetic acid, alpha-cyclopropylglycine, s-amino cyclopropyl acetic acid
PubChem CID 1501944
SMILES [NH3+][C@@H](C1CC1)C([O-])=O

Specifications

Color White
Beilstein 5728866
Solubility Information Soluble in water. Insoluble in ethanol.
IUPAC Name (2S)-2-azaniumyl-2-cyclopropylacetate
Quantity 250 mg
Formula Weight 115.13
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 2-Cyclopropyl-L-glycine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only