accessibility menu, dialog, popup

UserName

H3N(S)OO

CAS RN 49606-99-7

IUPAC Name: (2S)-2-azaniumyl-2-cyclopropylacetate
Synonyms: (2S)-2-ammonio-2-cyclopropylacetate
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: BUSBCPMSNBMUMT-BYPYZUCNSA-N
SMILES: [NH3+][C@@H](C1CC1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)