missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentylphenol, 98+%

Catalog No. p-4089606
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426510010 1 g Glass bottle
AC426510050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426510010 Supplier Thermo Scientific Chemicals Supplier No. 426510010

CAS: 1518-84-9 | C11H14O | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1518-84-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00009952
InChI Key JHEKSKQMOBLXQS-UHFFFAOYSA-N
Synonym phenol, 2-cyclopentyl, 2-cyclopentyl-phenol, o-cyclopentylphenol, o-cyclopentyl-phenol, 2-cyclopentyl phenol, acmc-20anxa, phenol, o-cyclopentyl, 2-cyclopentylphenol, o-cyclopentyl, 2-cyclopentylphenol, technical grade
PubChem CID 80285
IUPAC Name 2-cyclopentylphenol
SMILES OC1=CC=CC=C1C1CCCC1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 36°C
Color Colorless to Yellow
Boiling Point 148°C to 150°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 162.23
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Cyclopentylphenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-179-5

RUO – Research Use Only