missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentyloxypyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific™

$194.18 - $194.18

Chemical Identifiers

CAS 1073371-90-0
Molecular Formula C16H24BNO3
Molecular Weight (g/mol) 289.18
MDL Number MFCD07781170
InChI Key GZCZGFSEHAZEPS-UHFFFAOYSA-N
Synonym 2-cyclopentyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxypyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy pyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxy pyridine-3-boronic acid, pinacol ester, 2-cyclopentyloxy pyridin-3-yl boronic acid pinacol ester
PubChem CID 24208792
IUPAC Name 2-cyclopentyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(OC2CCCC2)N=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5011003
View Documents
Thermo Scientific Chemicals
H5011003
1 g
Each for $194.18
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1073371-90-0
289.18
GZCZGFSEHAZEPS-UHFFFAOYSA-N
24208792
CC1(C)OB(OC1(C)C)C1=C(OC2CCCC2)N=CC=C1
C16H24BNO3
MFCD07781170
2-cyclopentyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxypyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy pyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxy pyridine-3-boronic acid, pinacol ester, 2-cyclopentyloxy pyridin-3-yl boronic acid pinacol ester
2-cyclopentyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Specifications

1073371-90-0
MFCD07781170
2-cyclopentyloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxypyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy pyridine-3-boronic acid pinacol ester, 2-cyclopentyloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-cyclopentyloxy pyridine-3-boronic acid, pinacol ester, 2-cyclopentyloxy pyridin-3-yl boronic acid pinacol ester
CC1(C)OB(OC1(C)C)C1=C(OC2CCCC2)N=CC=C1
289.18
289.19
2-Cyclopentyloxypyridine-3-boronic acid pinacol ester
C16H24BNO3
1 g
GZCZGFSEHAZEPS-UHFFFAOYSA-N
2-cyclopentyloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
24208792
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'