missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentylethanol, 98+%

Catalog No. AAB2234406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2234406 5 g
AAB2234403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2234406 Supplier Thermo Scientific Chemicals Supplier No. B2234406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 766-00-7 | C7H14O | 114.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766-00-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00019301
InChI Key JEXQWCBPEWHFKC-UHFFFAOYSA-N
Synonym cyclopentaneethanol, 2-cyclopentylethan-1-ol, 2-cyclopentaneethanol, 2-hydroxyethylcyclopentane, 2-cyclopentyl-ethanol, 2-cyclopentyl ethanol, cyclopentylethanol, cyclopentane-ethanol, 2-cyclopentanethanol, acmc-1bgun
PubChem CID 69833
IUPAC Name 2-cyclopentylethanol
SMILES C1CCC(C1)CCO

Specifications

Density 0.92
Boiling Point 182°C
Flash Point 70°C (158°F)
Refractive Index 1.4585
Quantity 5 g
Beilstein 2069860
Formula Weight 114.19
Percent Purity ≥98%
Chemical Name or Material 2-Cyclopentylethanol
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 212-156-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only