accessibility menu, dialog, popup

UserName

OH

CAS RN 766-00-7

IUPAC Name: 2-cyclopentylethan-1-ol
Synonyms: 2-hydroxyethylcyclopentane
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: JEXQWCBPEWHFKC-UHFFFAOYSA-N
SMILES: OCCC1CCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)