missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentaneethanol 98.0+%, TCI America™

Catalog No. p-7124627
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08175ML 5 mL
C081725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08175ML Supplier TCI America Supplier No. C08175ML
Only null left

Chemical Identifiers

CAS 766-00-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00019301
InChI Key JEXQWCBPEWHFKC-UHFFFAOYSA-N
Synonym cyclopentaneethanol, 2-cyclopentylethan-1-ol, 2-cyclopentaneethanol, 2-hydroxyethylcyclopentane, 2-cyclopentyl-ethanol, 2-cyclopentyl ethanol, cyclopentylethanol, cyclopentane-ethanol, 2-cyclopentanethanol, acmc-1bgun
PubChem CID 69833
IUPAC Name 2-cyclopentylethanol
SMILES C1CCC(C1)CCO

Specifications

Color Yellow
Boiling Point 182°C
Quantity 5 mL
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclopentaneethanol