missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. p-7125830
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31011G 1 g
C31015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31011G Supplier TCI America Supplier No. C31011G

Chemical Identifiers

CAS 87100-15-0
Molecular Formula C12H23BO2
Molecular Weight (g/mol) 210.124
MDL Number MFCD04038749
InChI Key OUEVCDGYTKLNMJ-UHFFFAOYSA-N
Synonym cyclohexylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-cyclohexyl-4,4,5,5-tetramethyl, cyclohexylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane, pubchem7924, cyclohexylboronic acid, pinacol eser, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane
PubChem CID 3668596
IUPAC Name 2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2CCCCC2

Specifications

Color Colorless
Quantity 1 g
Formula Weight 210.12
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator