missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-ol 95.0+%, TCI America™

Catalog No. p-7124626
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08165ML 5 mL
C081625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08165ML Supplier TCI America Supplier No. C08165ML
Only null left

Chemical Identifiers

CAS 822-67-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001570
InChI Key PQANGXXSEABURG-UHFFFAOYSA-N
Synonym 2-cyclohexen-1-ol, cyclohex-2-enol, 2-cyclohexene-1-ol, 2-cyclohexenol, 3-hydroxycyclohexene, 1-cyclohexen-3-ol, cyclohexen-3-ol, cyclohex-2-ene-1-ol, 1,2,3,4-tetrahydrophenol, cyclohexen-3-ol van
PubChem CID 13198
IUPAC Name cyclohex-2-en-1-ol
SMILES C1CC=CC(C1)O

Specifications

Color Yellow
Boiling Point 166°C
Quantity 5 mL
UN Number 1987
Formula Weight 98.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclohexen-1-ol