accessibility menu, dialog, popup

UserName

CAS RN 822-67-3

IUPAC Name: cyclohex-2-en-1-ol
Synonyms: 2-cyclohexen-1-ol
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: PQANGXXSEABURG-UHFFFAOYNA-N
SMILES: OC1CCCC=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)