missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-ol, 95%

Catalog No. AC111230010
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111230010 1 g Glass bottle
AC111230050 5 g Glass bottle
AC111230100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111230010 Supplier Thermo Scientific Chemicals Supplier No. 111230010

CAS: 822-67-3 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 822-67-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00001570
InChI Key PQANGXXSEABURG-UHFFFAOYSA-N
Synonym 2-cyclohexen-1-ol, cyclohex-2-enol, 2-cyclohexene-1-ol, 2-cyclohexenol, 3-hydroxycyclohexene, 1-cyclohexen-3-ol, cyclohexen-3-ol, cyclohex-2-ene-1-ol, 1,2,3,4-tetrahydrophenol, cyclohexen-3-ol van
PubChem CID 13198
IUPAC Name cyclohex-2-en-1-ol
SMILES C1CC=CC(C1)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.9840g/mL
Boiling Point 164.0°C to 166.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4855
Linear Formula C6H9OH
Quantity 1 g
Beilstein 06,II,6
Specific Gravity 0.984
Formula Weight 98.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Cyclohexen-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapour.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-501-3

RUO – Research Use Only