missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohepten-1-one 85.0+%, TCI America™

Catalog No. C31801G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31801G 1 g
C31805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31801G Supplier TCI America Supplier No. C31801G

Chemical Identifiers

CAS 1121-66-0
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00004157
InChI Key WZCRDVTWUYLPTR-UHFFFAOYSA-N
Synonym 2-cyclohepten-1-one, cyclohept-2-enone, tropilene, 2-cycloheptenone, cycloheptenone, z-cyclohept-2-enone, cyclohept-2-eneone, cyclohept-en-1-one, acmc-1bphl, 2-cyclohepten-1-one, z
PubChem CID 70723
IUPAC Name cyclohept-2-en-1-one
SMILES C1CCC(=O)C=CC1

Specifications

Color Yellow
Boiling Point 183°C
Quantity 1 g
Formula Weight 110.16
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclohepten-1-one
TSCA No
Recommended Storage Refrigerator