missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohepten-1-one, 90%

Catalog No. AC171070010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171070010 1 g
AC171070050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC171070010 Supplier Thermo Scientific Chemicals Supplier No. 171070010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1121-66-0 | C7H10O | 110.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1121-66-0
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
InChI Key WZCRDVTWUYLPTR-UHFFFAOYSA-N
Synonym 2-cyclohepten-1-one, cyclohept-2-enone, tropilene, 2-cycloheptenone, cycloheptenone, z-cyclohept-2-enone, cyclohept-2-eneone, cyclohept-en-1-one, acmc-1bphl, 2-cyclohepten-1-one, z
PubChem CID 70723
IUPAC Name cyclohept-2-en-1-one
SMILES C1CCC(=O)C=CC1

Specifications

Density 0.9900g/mL
Boiling Point 189.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 89% min. (GC)
Packaging Glass Bottle
Refractive Index 1.495
Linear Formula C7H10(=O)
Quantity 1 g
Specific Gravity 0.99
Formula Weight 110.16
Percent Purity 90%
Chemical Name or Material 2-Cyclohepten-1-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.
EINECSNumber 214-334-1

RUO – Research Use Only