missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanothiazole, 95%

CAS: 1452-16-0 | C4H2N2S | 110.134 g/mol

Supplier:  Thermo Scientific Chemicals H6342506

 View more versions of this product

Catalog No. AAH6342506

Only null left
Explore available promotions

Description

Description

2-Cyanothiazole is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanothiazole is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-Cyanothiazole
C4H2N2S
5 g
2-cyanothiazole, thiazole-2-carbonitrile, 2-thiazolecarbonitrile, 2-cyanothiazol
ZIRGWUZHKJDYKT-UHFFFAOYSA-N
1,3-thiazole-2-carbonitrile
15111480
95%
1452-16-0
MFCD09702472
UN3439
Slightly soluble in water.
C1=CSC(=N1)C#N
110.134
110.14
Solid

Chemical Identifiers

1452-16-0
110.134
ZIRGWUZHKJDYKT-UHFFFAOYSA-N
15111480
C1=CSC(=N1)C#N
C4H2N2S
MFCD09702472
2-cyanothiazole, thiazole-2-carbonitrile, 2-thiazolecarbonitrile, 2-cyanothiazol
1,3-thiazole-2-carbonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H371-H302-H312-H332-H315-H319
May cause damage to organs.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P309+P311-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H371

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only