missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanothiazole, 95%

Catalog No. p-7054965
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63425MD 250 mg
AAH6342503 1 g
AAH6342506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63425MD Supplier Thermo Scientific Chemicals Supplier No. H63425MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1452-16-0 | C4H2N2S | 110.134 g/mol

2-Cyanothiazole is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanothiazole is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1452-16-0
Molecular Formula C4H2N2S
Molecular Weight (g/mol) 110.134
MDL Number MFCD09702472
InChI Key ZIRGWUZHKJDYKT-UHFFFAOYSA-N
Synonym 2-cyanothiazole, thiazole-2-carbonitrile, 2-thiazolecarbonitrile, 2-cyanothiazol
PubChem CID 15111480
IUPAC Name 1,3-thiazole-2-carbonitrile
SMILES C1=CSC(=N1)C#N

Specifications

Quantity 250 mg
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 110.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Cyanothiazole
Hazard Category H302+H312+H332-H315-H319-H371
Hazard Statement GHS H Statement
H371-H302-H312-H332-H315-H319
May cause damage to organs.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P309+P311-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only