missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanopyrimidine 98.0+%, TCI America™

Catalog No. c15181g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
5 g
Catalog No. Quantity
C15181G 1 g
C15185G 5 g
C151825G 25 g
3 options

Catalog No. C15181G

Supplier: TCI America C15181G

Only null left

Chemical Identifiers

CAS 14080-23-0
Molecular Formula C5H3N3
Molecular Weight (g/mol) 105.10
MDL Number MFCD00160513
InChI Key IIHQNAXFIODVDU-UHFFFAOYSA-N
Synonym 2-cyanopyrimidine, 2-pyrimidinecarbonitrile, 2-cyano-pyrimidine, 2-cyano pyrimidine, cyanopyrimidine, provastatin, pyrimidinecarbonitrile, zlchem 283, pubchem7042, pyrimidin-2-carbonitrile
PubChem CID 2757979
IUPAC Name pyrimidine-2-carbonitrile
SMILES N#CC1=NC=CC=N1

Specifications

Melting Point 41°C
Quantity 1 g
Formula Weight 105.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanopyrimidine