missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyanophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$74.56 - $74.56
Chemical Identifiers
| CAS | 138642-62-3 |
|---|---|
| Molecular Formula | C7H6BNO2 |
| Molecular Weight (g/mol) | 146.94 |
| MDL Number | MFCD01632208 |
| InChI Key | NPLZNDDFVCGRAG-UHFFFAOYSA-N |
| PubChem CID | 2734610 |
| IUPAC Name | (2-cyanophenyl)boronic acid |
| SMILES | OB(O)C1=CC=CC=C1C#N |
Chemical Identifiers
| 138642-62-3 | |
| 146.94 | |
| NPLZNDDFVCGRAG-UHFFFAOYSA-N | |
| (2-cyanophenyl)boronic acid |
| C7H6BNO2 | |
| MFCD01632208 | |
| 2734610 | |
| OB(O)C1=CC=CC=C1C#N |
Specifications
| 138642-62-3 | |
| White | |
| MFCD01632208 | |
| 2-cyanophenyl boronic acid, 2-cyanobenzeneboronic acid, 2-cyanophenylboric acid, 2-cyanophenylboronicacid, 2-dihydroxyboranyl benzonitrile, boronic acid, b-2-cyanophenyl, 2-boronobenzonitrile, 2-cyanobenzene boronic acid, 2-cyano-phenyl-boronic acid, boronic acid, cyanophenyl | |
| OB(O)C1=CC=CC=C1C#N | |
| 146.94 | |
| 146.94 | |
| 2-Cyanophenylboronic Acid (contains varying amounts of Anhydride) |
| 240°C | |
| C7H6BNO2 | |
| 1 g | |
| NPLZNDDFVCGRAG-UHFFFAOYSA-N | |
| (2-cyanophenyl)boronic acid | |
| 2734610 | |
| Crystalline Powder |
Safety and Handling
TSCA : No