missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanophenol 98.0+%, TCI America™

Catalog No. p-7124758
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C10455G 5 g
C104525G 25 g
2 options

Catalog No. C10455G

Supplier: TCI America C10455G

Only null left

Chemical Identifiers

CAS 611-20-1
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00002145
InChI Key CHZCERSEMVWNHL-UHFFFAOYSA-N
Synonym 2-cyanophenol, o-cyanophenol, salicylonitrile, o-hydroxybenzonitrile, benzonitrile, 2-hydroxy, salicylnitrile, benzonitrile, hydroxy, benzonitrile, o-hydroxy, o-hydoxybenzonitrile, 2-hydroxy-benzonitrile
PubChem CID 11907
IUPAC Name 2-hydroxybenzonitrile
SMILES C1=CC=C(C(=C1)C#N)O

Specifications

Melting Point 97°C
Boiling Point 149°C
Quantity 5 g
Formula Weight 119.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanophenol