missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethyl N,N,N',N'-Tetraisopropylphosphordiamidite 95.0+%, TCI America™

Catalog No. C22281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22281G 1 g
C22285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22281G Supplier TCI America Supplier No. C22281G

Chemical Identifiers

CAS 102691-36-1
Molecular Formula C15H32N3OP
Molecular Weight (g/mol) 301.415
MDL Number MFCD00012213
InChI Key RKVHNYJPIXOHRW-UHFFFAOYSA-N
Synonym 3-bis diisopropylamino phosphino oxy propanenitrile, bis diisopropylamino 2-cyanoethoxy phosphine, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl-n,n,n',n'-tetraisopropylphosphorodiamidite, 2-cyanoethyl n,n,n',n'-tetraisopropylphosphordiamidite, phosphorodiamidous acid, tetrakis 1-methylethyl-, 2-cyanoethyl ester, 2-cyanoethyl n,n,n',n'-tetraisopropyl-phosphordiamidite, 2-cyanoethyl tetraisopropylphosphorodiamidite, cetipd, 3-bis diisopropylamino phosphinooxy propanenitrile
PubChem CID 128153
IUPAC Name 3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile
SMILES CC(C)N(C(C)C)P(N(C(C)C)C(C)C)OCCC#N

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Formula Weight 301.41
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Cyanoethyl N,N,N′,N′-Tetraisopropylphosphordiamidite
TSCA No
Recommended Storage Refrigerator