accessibility menu, dialog, popup

UserName

CAS RN 102691-36-1

IUPAC Name: 3-({bis[bis(propan-2-yl)amino]phosphanyl}oxy)propanenitrile
Synonyms: 3-{[bis(diisopropylamino)phosphanyl]oxy}propanenitrile
Molecular Weight (g/mol): 301.42
Molecular Formula: C15H32N3OP
InChi Key: RKVHNYJPIXOHRW-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)P(OCCC#N)N(C(C)C)C(C)C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)