missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoethyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. p-7125216
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19745G 5 g
C197425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19745G Supplier TCI America Supplier No. C19745G

Chemical Identifiers

CAS 106-71-8
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.13
MDL Number MFCD00013823
InChI Key AEPWOCLBLLCOGZ-UHFFFAOYSA-N
Synonym Acrylic Acid 2-Cyanoethyl Ester, 3-Acryloyloxypropionitrile
PubChem CID 7825
IUPAC Name 2-cyanoethyl prop-2-enoate
SMILES C=CC(=O)OCCC#N

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
UN Number 3276
Formula Weight 125.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyanoethyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-3416
RTECSNumber AT1500000
TSCA Yes