accessibility menu, dialog, popup

UserName

H2COON

CAS RN 106-71-8

IUPAC Name: 2-cyanoethyl prop-2-enoate
Synonyms: 2-cyanoethyl acrylate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: AEPWOCLBLLCOGZ-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results