missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzyl Bromide 98.0+%, TCI America™

Catalog No. C155625G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C155625G 25 g
C155610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C155625G Supplier TCI America Supplier No. C155625G

Chemical Identifiers

CAS 22115-41-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001794
InChI Key QGXNHCXKWFNKCG-UHFFFAOYSA-N
Synonym 2-bromomethyl benzonitrile, 2-cyanobenzyl bromide, alpha-bromo-o-tolunitrile, o-cyanobenzyl bromide, a-bromo-o-tolunitrile, alpha bromo-o-tolunitrile, 2 bromomethyl benzonitrile, 2-bromomethyl-benzonitrile, 2-cyanophenyl methyl bromide, benzonitrile, 2-bromomethyl
PubChem CID 89599
IUPAC Name 2-(bromomethyl)benzonitrile
SMILES BrCC1=CC=CC=C1C#N

Specifications

Melting Point 72°C
Boiling Point 111°C
Quantity 25 g
UN Number 3261
Formula Weight 196.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanobenzyl Bromide