missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanoacetamide 98.0+%, TCI America™

Catalog No. p-7124419
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C043725G 25 g
C0437500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C043725G Supplier TCI America Supplier No. C043725G
Only null left

Chemical Identifiers

CAS 107-91-5
Molecular Formula C3H4N2O
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008024
InChI Key DGJMPUGMZIKDRO-UHFFFAOYSA-N
Synonym cyanoacetamide, acetamide, 2-cyano, cyanacetamide, malonamide nitrile, malonamonitrile, nitrilomalonamide, cyanoiminoacetic acid, 3-nitrilo-propionamide, propionamide, 3-nitrilo, usaf kf-14
PubChem CID 7898
IUPAC Name 2-cyanoacetamide
SMILES NC(=O)CC#N

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 25 g
Formula Weight 84.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyanoacetamide
EINECSNumber (2)-1550
RTECSNumber AB5950000
TSCA Yes