missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-6-fluoropyridine, 95%

Catalog No. AAH27600MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27600MD 250 mg
AAH2760003 1 g
AAH2760006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH27600MD Supplier Thermo Scientific Chemicals Supplier No. H27600MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3939-15-9 | C6H3FN2 | 122.10 g/mol

It is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3939-15-9
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.10
MDL Number MFCD09263323
InChI Key NVOLTPVZQXTZCW-UHFFFAOYSA-N
Synonym 2-cyano-6-fluoropyridine, 6-fluoropicolinonitrile, 2-pyridinecarbonitrile, 6-fluoro, 6-fluoro-2-cyanopyridine;2-fluoro-6-cyanopyridine;6-fluoropicolinonitrile, pubchem4017, acmc-1cj0q, 2-pyridinecarbonitrile,6-fluoro, 6-fluoro-pyridine-2-carbonitrile, 2-fluoropyridine-6-carbonitrile, 6-fluoro-2-pyridinecarbonitrile
PubChem CID 14296369
IUPAC Name 6-fluoropyridine-2-carbonitrile
SMILES FC1=CC=CC(=N1)C#N

Specifications

Melting Point 32°C to 35°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 122.1
Percent Purity 95%
Chemical Name or Material 2-Cyano-6-fluoropyridine
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only