CAS RN 3939-15-9
IUPAC Name:
6-fluoropyridine-2-carbonitrile
Synonyms:
6-fluoropyridine-2-carbonitrile
Molecular Weight (g/mol):
122.1
Molecular Formula:
C6H3FN2
InChi Key:
NVOLTPVZQXTZCW-UHFFFAOYSA-N
SMILES:
FC1=CC=CC(=N1)C#N
1
–
1
of
1
results
1
Promotions Available
2-Cyano-6-fluoropyridine, 95%
2-Cyano-6-fluoropyridine, 95%
CAS: 3939-15-9 Molecular Formula: C6H3FN2 Molecular Weight (g/mol): 122.10 MDL Number: MFCD09263323 InChI Key: NVOLTPVZQXTZCW-UHFFFAOYSA-N Synonym: 2-cyano-6-fluoropyridine,6-fluoropicolinonitrile,2-pyridinecarbonitrile, 6-fluoro,6-fluoro-2-cyanopyridine;2-fluoro-6-cyanopyridine;6-fluoropicolinonitrile,pubchem4017,acmc-1cj0q,2-pyridinecarbonitrile,6-fluoro,6-fluoro-pyridine-2-carbonitrile,2-fluoropyridine-6-carbonitrile,6-fluoro-2-pyridinecarbonitrile PubChem CID: 14296369 IUPAC Name: 6-fluoropyridine-2-carbonitrile SMILES: FC1=CC=CC(=N1)C#N