missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-5-methylpyridine 98.0+%, TCI America™

Catalog No. p-7125607
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27061G 1 g
C27065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27061G Supplier TCI America Supplier No. C27061G

Chemical Identifiers

CAS 1620-77-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD06200830
InChI Key LIEQVZZZYLHNRH-UHFFFAOYSA-N
Synonym 2-cyano-5-methylpyridine, 5-methylpicolinonitrile, 5-methyl-pyridine-2-carbonitrile, 5-methyl-2-pyridinecarbonitrile, 6-cyano-3-picoline, 2-pyridinecarbonitrile, 5-methyl, 2-cyano-5-methylpyridine 2-cyano-5-picoline, 5-methypicolinonitrile, pubchem12978, 5-methyl picolinonitrile
PubChem CID 74183
IUPAC Name 5-methylpyridine-2-carbonitrile
SMILES CC1=CC=C(N=C1)C#N

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 140°C
Quantity 1 g
UN Number 3439
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Cyano-5-methylpyridine