missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-methoxybenzeneboronic acid, 95%

Catalog No. AAH61856MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61856MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH61856MC Supplier Thermo Scientific Chemicals Supplier No. H61856MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1164100-84-8 | C8H8BNO3 | 176.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1164100-84-8
Molecular Formula C8H8BNO3
Molecular Weight (g/mol) 176.97
MDL Number MFCD10696671
InChI Key GRHINHWGIHWEPE-UHFFFAOYSA-N
Synonym 2-cyano-3-methoxyphenyl boronic acid, acmc-2099sp, 2-cyano-3-methoxyphenyl boronicacid
PubChem CID 53216734
IUPAC Name (2-cyano-3-methoxyphenyl)boronic acid
SMILES COC1=CC=CC(B(O)O)=C1C#N

Specifications

Quantity 100 mg
Formula Weight 176.97
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Cyano-3-methoxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only