accessibility menu, dialog, popup

UserName

BOCH3NHOOH

CAS RN 1164100-84-8

IUPAC Name: (2-cyano-3-methoxyphenyl)boronic acid
Synonyms: 2-cyano-3-methoxyphenylboronic acid
Molecular Weight (g/mol): 176.97
Molecular Formula: C8H8BNO3
InChi Key: GRHINHWGIHWEPE-UHFFFAOYSA-N
SMILES: COC1=CC=CC(B(O)O)=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results