missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-iodopyridine, 95%

Catalog No. AAH26963MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26963MD 250 mg
AAH2696303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26963MD Supplier Thermo Scientific Chemicals Supplier No. H26963MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 827616-52-4 | C6H3IN2 | 230.008 g/mol

Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare Grignard reagents which are widely used in organic synthesis.2-Cyano-3-iodopyridine are used in coupling reactions. Has wide application in the pharmaceutical as well as chemical industries.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Chemical Identifiers

CAS 827616-52-4
Molecular Formula C6H3IN2
Molecular Weight (g/mol) 230.008
MDL Number MFCD09037451
InChI Key MZZGCCSFZCRVAF-UHFFFAOYSA-N
Synonym 3-iodopicolinonitrile, 2-cyano-3-iodopyridine, 2-pyridinecarbonitrile,3-iodo, pubchem15743
PubChem CID 12180890
IUPAC Name 3-iodopyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)I

Specifications

Melting Point 98°C to 100°C
Quantity 250 mg
UN Number UN3439
Sensitivity Light sensitive
Formula Weight 230.01
Percent Purity 95%
Chemical Name or Material 2-Cyano-3-iodopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only